Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01747277_m1
          See other ITGB6 GE Assays ›
          Gene Symbol:
          Itgb6
          Gene Name:
          integrin, beta 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 46652624 - 46775363 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01747277_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itgb6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004263.1 NP_001004263.1 15 - 16 2330 82
          XM_006234232.1 XP_006234294.1 17 - 18 2673 82
          XM_006234233.2 XP_006234295.1 15 - 16 2579 82
          XM_008761845.1 XP_008760067.1 17 - 18 2500 82
          XM_008761846.1 XP_008760068.1 15 - 16 2477 82
          GenBank mRNA BC079069.1 - 15 - 16 2330 7121642 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003178.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Itgb6

          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          cell-matrix adhesion
          integrin-mediated signaling pathway
          multicellular organism development
          biological_process
          molecular_function
          receptor activity
          integrin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P755326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115477 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91370 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157827 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124034 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170710 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133992 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97170 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P755326 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P755326 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P755326 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P755326 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78935 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71437 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61170 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7812 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23159 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96764 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10845598 - -
          Affymetrix Rat Gene 1.0 ST Array 10845594 - -
          Affymetrix Rat Gene 1.0 ST Array 10845593 - -
          Affymetrix Rat Gene 1.0 ST Array 10845600 - -
          Affymetrix Rat Gene 1.0 ST Array 10845588 - -
          Affymetrix Rat Gene 1.0 ST Array 10845590 - -
          Affymetrix Rat Gene 1.0 ST Array 10845591 - -
          Affymetrix Rat Gene 1.0 ST Array 10845595 - -
          Affymetrix Rat Gene 1.0 ST Array 10845597 - -
          Affymetrix Rat Gene 1.0 ST Array 10845589 - -
          Affymetrix Rat Gene 1.0 ST Array 10845592 - -
          Affymetrix Rat Gene 1.0 ST Array 10845599 - -
          Affymetrix Rat Gene 1.0 ST Array 10845601 - -
          Affymetrix Rat Gene 1.0 ST Array 10845596 - -
          Affymetrix Rat Gene 1.0 ST Array 10845602 - -
          Affymetrix Rat Gene 1.1 ST Array 10845587 - -
          Rat Genome 230 2.0 Array 1382439_at - -
          Rat Genome 230 2.0 Array 1382439_PM_at - -
          Affymetrix HT Rat Focus Array 1382439_PM_at - -
          Affymetrix HT Rat Focus Array 1382439_at - -
          Affymetrix Rat Clariom S Assay PSR0300024618.rn.1 TC0300003178.rn.1 RTA TC
          Rat Expression Set 230 1382439_PM_at - -
          Rat Expression Set 230 1382439_at - -
          Affymetrix Rat Gene 1.0 ST Array 10845587 - -
          Rat Genome U34 Set rc_AI171816_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20989619 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357529 - -
          RatRef-12 Expression BeadChip ILMN_66282 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline