Search
Search

Tspan18| Gene Symbol: Tspan18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107750.1 | NP_001101220.1 | 8 - 9 | 1118 | 64 | |
| XM_006234573.2 | XP_006234635.1 | 8 - 9 | 1049 | 64 | ||
| XM_006234574.2 | XP_006234636.1 | 9 - 10 | 1191 | 64 | ||
| XM_006234575.2 | XP_006234637.1 | 7 - 8 | 969 | 64 | ||
| XM_008762027.1 | XP_008760249.1 | 7 - 8 | 869 | 64 | ||
Set Membership: |
|