Search
Search
| Gene Symbol: Ankrd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192012.1 | NP_001178941.1 | 3 - 4 | 535 | 114 | |
| XM_006236119.2 | XP_006236181.1 | 3 - 4 | 1765 | 114 | ||
| XM_006236120.2 | XP_006236182.1 | 3 - 4 | 1768 | 114 | ||
| XM_006236121.2 | XP_006236183.1 | 3 - 4 | 1768 | 114 | ||
Set Membership: |
|