Search
Search
| Gene Symbol: Lrrc39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109637.1 | NP_001103107.1 | 7 - 8 | 1047 | 70 | |
| XM_006233231.2 | XP_006233293.1 | 7 - 8 | 1034 | 70 | ||
| GenBank mRNA | BC158606.1 | - | 7 - 8 | 1003 | 70 | |
| FQ215425.1 | - | 7 - 8 | 988 | 70 | ||
Set Membership: |
|