Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01748499_m1
          See other DKK2 GE Assays ›
          Gene Symbol:
          Dkk2
          Gene Name:
          dickkopf WNT signaling pathway inhibitor 2
          Gene Aliases:
          Dkk4
          Chromosome Location:
          Chr.2: 237148918 - 237240883 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01748499_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dkk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106472.1 NP_001099942.1 3 - 4 1277 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002227.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          multicellular organism development
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P103414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353422 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94089 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26329 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81767 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P103414 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P103414 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P103414 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P623856 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P293033 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53495 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10819077 - -
          Affymetrix Rat Gene 1.0 ST Array 10819075 - -
          Affymetrix Rat Gene 1.0 ST Array 10819076 - -
          Affymetrix Rat Gene 1.0 ST Array 10819079 - -
          Affymetrix Rat Gene 1.1 ST Array 10819074 - -
          Rat Genome 230 2.0 Array 1397684_at - -
          Rat Genome 230 2.0 Array 1385444_at - -
          Rat Genome 230 2.0 Array 1397684_PM_at - -
          Rat Genome 230 2.0 Array 1385444_PM_at - -
          Affymetrix HT Rat Focus Array 1397684_PM_at - -
          Affymetrix HT Rat Focus Array 1397684_at - -
          Affymetrix HT Rat Focus Array 1385444_PM_at - -
          Affymetrix HT Rat Focus Array 1385444_at - -
          Affymetrix Rat Clariom S Assay PSR0200015333.rn.1 TC0200002227.rn.1 RTA TC
          Rat Expression Set 230 1385444_at - -
          Rat Expression Set 230 1385444_PM_at - -
          Rat Expression Set 230 1397684_at - -
          Rat Expression Set 230 1397684_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10819074 - -
          Rat Genome U34 Set rc_AI071994_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20796035 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363501 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline