Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01748618_m1
          See other LOC691931 GE Assays ›
          Gene Symbol:
          LOC691931
          Gene Name:
          hypothetical protein LOC691931
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 232214331 - 232245319 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01748618_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC691931
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109651.1 NP_001103121.1 1 - 2 253 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004665.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P164926 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P042402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509450 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158635 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42606 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88490 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042402 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164926 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164926 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042402 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P164926 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042402 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36017 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10826704 - -
          Affymetrix Rat Gene 1.0 ST Array 10826706 - -
          Affymetrix Rat Gene 1.1 ST Array 10826703 - -
          Rat Genome 230 2.0 Array 1376618_PM_at - -
          Rat Genome 230 2.0 Array 1376618_at - -
          Affymetrix Rat Clariom S Assay PSR0200032464.rn.1 TC0200004665.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200032462.rn.1 TC0200004665.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200032463.rn.1 TC0200004665.rn.1 RTA TC
          Rat Expression Set 230 1376618_PM_at - -
          Rat Expression Set 230 1376618_at - -
          Affymetrix Rat Gene 1.0 ST Array 10826703 - -
          Rat Genome U34 Set rc_AI236218_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21875253 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline