Search
Search
| Gene Symbol: Stk38l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083336.1 | NP_001076805.1 | 10 - 11 | 987 | 67 | |
| XM_008763431.1 | XP_008761653.1 | 9 - 10 | 1884 | 67 | ||
| GenBank mRNA | BC160812.1 | - | 8 - 9 | 869 | 7460150 | 67 |
| EF444939.1 | - | 10 - 11 | 987 | 67 | ||
Set Membership: |
|