Search
Search

Clec12a| Gene Symbol: Clec12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134716.1 | NP_001128188.1 | 579 | 74 | ||
| GenBank mRNA | BC168186.1 | - | 579 | 7100019 | 74 | |
| GU357484.1 | - | 378 | 74 | |||
Set Membership: |
|