Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01749021_g1
          See other KLRA1 GE Assays ›
          Gene Symbol:
          Klra1
          Gene Name:
          killer cell lectin-like receptor, subfamily A, member 1
          Gene Aliases:
          Klra2, Ly49i8
          Chromosome Location:
          Chr.4: 165426365 - 165460142 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01749021_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klra1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009486.1 NP_001009486.1 8 - 9 998 115
          XM_008763404.1 XP_008761626.1 7 - 8 975 115
          GenBank mRNA AY649835.1 - 8 - 9 998 115
          BC085917.1 - 961 7108565 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003888.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klra1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158948 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72829 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147860 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052265 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052265 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052265 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P652293 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11225 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96567 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40354 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95330 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43696 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866256 - -
          Affymetrix Rat Gene 1.0 ST Array 10866262 - -
          Affymetrix Rat Gene 1.0 ST Array 10866264 - -
          Affymetrix Rat Gene 1.0 ST Array 10866258 - -
          Affymetrix Rat Gene 1.0 ST Array 10866259 - -
          Affymetrix Rat Gene 1.0 ST Array 10866260 - -
          Affymetrix Rat Gene 1.0 ST Array 10866257 - -
          Affymetrix Rat Gene 1.0 ST Array 10866253 - -
          Affymetrix Rat Gene 1.0 ST Array 10866254 - -
          Affymetrix Rat Gene 1.0 ST Array 10866261 - -
          Affymetrix Rat Gene 1.0 ST Array 10866263 - -
          Affymetrix Rat Gene 1.0 ST Array 10866255 - -
          Affymetrix Rat Gene 1.1 ST Array 10866252 - -
          Affymetrix Rat Clariom S Assay PSR0400027773.rn.1 TC0400003888.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027776.rn.1 TC0400003888.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027778.rn.1 TC0400003888.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027777.rn.1 TC0400003888.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027772.rn.1 TC0400003888.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027774.rn.1 TC0400003888.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10866252 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20721916 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367927 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0