Search
Search
| Gene Symbol: Eps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237583.2 | XP_006237645.1 | 15 - 16 | 2443 | 61 | |
| XM_008763452.1 | XP_008761674.1 | 15 - 16 | 1727 | 61 | ||
| XM_232499.8 | XP_232499.6 | 14 - 15 | 2026 | 61 | ||
Set Membership: |
|