Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01749101_s1
          See other LOC500354 GE Assays ›
          Gene Symbol:
          LOC500354
          Gene Name:
          similar to C030030A07Rik protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 170810582 - 170822358 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01749101_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC500354
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037797.1 NP_001032886.1 2 818 79
          XM_006237568.2 XP_006237630.1 2 807 79
          XM_008763417.1 XP_008761639.1 2 878 79
          GenBank mRNA BC098929.1 - 2 818 7133030 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC500354

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P153202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220292 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124232 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162324 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P153202 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P153202 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P153202 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P636936 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66933 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18426 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10859336 - -
          Affymetrix Rat Gene 1.1 ST Array 10859335 - -
          Affymetrix Rat Clariom S Assay PSR0400014111.rn.1 TC0400001962.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400014110.rn.1 TC0400001962.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400014112.rn.1 TC0400001962.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10859335 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22345002 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354628 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline