Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01749108_g1
          See other LOC502894 GE Assays ›
          Gene Symbol:
          LOC502894
          Gene Name:
          hypothetical protein LOC502894
          Gene Aliases:
          Tigar
          Chromosome Location:
          Chr.4: 159635145 - 159654104 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01749108_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC502894
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025064.1 NP_001020235.1 4 - 5 430 80
          XM_006237501.2 XP_006237563.1 5 - 6 509 80
          GenBank mRNA BC093610.1 - 4 - 5 430 7319640 80
          FQ217296.1 - 5 - 6 471 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC502894

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s)

          response to xenobiotic stimulus
          response to gamma radiation
          positive regulation of cardiac muscle cell apoptotic process
          intestinal epithelial cell development
          negative regulation of macromitophagy
          negative regulation of reactive oxygen species metabolic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P675139 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P063543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349379 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75037 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P675139 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063543 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063543 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P675139 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P675139 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P063543 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33229 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91294 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101599 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10865779 - -
          Affymetrix Rat Gene 1.0 ST Array 10884923 - -
          Affymetrix Rat Gene 1.0 ST Array 10865781 - -
          Affymetrix Rat Gene 1.0 ST Array 10865780 - -
          Affymetrix Rat Gene 1.0 ST Array 10884922 - -
          Affymetrix Rat Gene 1.0 ST Array 10865778 - -
          Affymetrix Rat Gene 1.1 ST Array 10884921 - -
          Affymetrix Rat Gene 1.1 ST Array 10865777 - -
          Rat Genome 230 2.0 Array 1374777_PM_at - -
          Rat Genome 230 2.0 Array 1374777_at - -
          Affymetrix HT Rat Focus Array 1374777_PM_at - -
          Affymetrix HT Rat Focus Array 1374777_at - -
          Affymetrix Rat Clariom S Assay PSR0400026960.rn.1 TC0400003790.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026961.rn.1 TC0400003790.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026962.rn.1 TC0400003790.rn.1 RTA TC
          Rat Expression Set 230 1374777_PM_at - -
          Rat Expression Set 230 1374777_at - -
          Affymetrix Rat Gene 1.0 ST Array 10865777 - -
          Rat Genome U34 Set rc_AI233024_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22381705 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374668 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline