Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01749180_m1
          See other MCMDC2 GE Assays ›
          Gene Symbol:
          Mcmdc2
          Gene Name:
          minichromosome maintenance domain containing 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 8973882 - 9038662 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01749180_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mcmdc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024340.1 NP_001019511.2 11 - 12 1490 62
          XM_006237781.2 XP_006237843.1 10 - 11 4316 62
          XM_006237782.2 XP_006237844.1 11 - 12 2119 62
          XM_006237784.2 XP_006237846.1 11 - 12 3871 62
          XM_006237785.2 XP_006237847.1 10 - 11 4303 62
          XM_006237787.2 XP_006237849.1 10 - 11 4315 62
          XM_006237788.2 XP_006237850.1 10 - 11 4313 62
          XM_006237789.2 XP_006237851.1 10 - 11 4313 62
          GenBank mRNA BC083882.1 - 11 - 12 1490 7131459 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002183.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mcmdc2

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          DNA binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P000381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280077 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49705 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107904 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81250 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2571 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000381 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000381 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P000381 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P806521 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66983 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76066 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3888 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10875105 - -
          Affymetrix Rat Gene 1.0 ST Array 10875106 - -
          Affymetrix Rat Gene 1.0 ST Array 10875103 - -
          Affymetrix Rat Gene 1.0 ST Array 10875107 - -
          Affymetrix Rat Gene 1.0 ST Array 10875114 - -
          Affymetrix Rat Gene 1.0 ST Array 10875110 - -
          Affymetrix Rat Gene 1.0 ST Array 10875109 - -
          Affymetrix Rat Gene 1.0 ST Array 10875113 - -
          Affymetrix Rat Gene 1.0 ST Array 10875104 - -
          Affymetrix Rat Gene 1.0 ST Array 10875108 - -
          Affymetrix Rat Gene 1.0 ST Array 10875111 - -
          Affymetrix Rat Gene 1.0 ST Array 10875115 - -
          Affymetrix Rat Gene 1.0 ST Array 10875112 - -
          Affymetrix Rat Gene 1.0 ST Array 10875116 - -
          Affymetrix Rat Gene 1.1 ST Array 10875102 - -
          Affymetrix Rat Clariom S Assay PSR0500016652.rn.1 TC0500002183.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016653.rn.1 TC0500002183.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016655.rn.1 TC0500002183.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016656.rn.1 TC0500002183.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016657.rn.1 TC0500002183.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10875102 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22061099 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349039 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline