Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01749291_m1
          See other TERF1 GE Assays ›
          Gene Symbol:
          Terf1
          Gene Name:
          telomeric repeat binding factor (NIMA-interacting) 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 2853455 - 2882728 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01749291_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Terf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012464.1 NP_001012482.1 9 - 10 1110 86
          XM_006237729.2 XP_006237791.1 8 - 9 1083 86
          GenBank mRNA BC089888.1 - 9 - 10 1110 7374057 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002133.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Terf1

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          obsolete age-dependent telomere shortening
          telomere maintenance via telomerase
          mitotic spindle assembly checkpoint
          negative regulation of DNA replication
          obsolete telomere maintenance via telomere shortening
          positive regulation of microtubule polymerization
          telomeric loop formation
          negative regulation of telomere maintenance via telomerase
          response to drug
          positive regulation of apoptotic process
          positive regulation of mitotic nuclear division
          positive regulation of mitotic cell cycle
          protein homooligomerization
          negative regulation of telomerase activity
          molecular_function
          chromatin binding
          double-stranded telomeric DNA binding
          microtubule binding
          DNA binding, bending
          telomeric DNA binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P267529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16743 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P267529 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P267529 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P267529 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20877 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P267529 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17660 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100349 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86558 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10874950 - -
          Affymetrix Rat Gene 1.0 ST Array 10874949 - -
          Affymetrix Rat Gene 1.0 ST Array 10874946 - -
          Affymetrix Rat Gene 1.0 ST Array 10874948 - -
          Affymetrix Rat Gene 1.0 ST Array 10874941 - -
          Affymetrix Rat Gene 1.0 ST Array 10874944 - -
          Affymetrix Rat Gene 1.0 ST Array 10874945 - -
          Affymetrix Rat Gene 1.0 ST Array 10874947 - -
          Affymetrix Rat Gene 1.0 ST Array 10874943 - -
          Affymetrix Rat Gene 1.0 ST Array 10874942 - -
          Affymetrix Rat Gene 1.0 ST Array 10874951 - -
          Affymetrix Rat Gene 1.1 ST Array 10874940 - -
          Rat Genome 230 2.0 Array 1394854_PM_at - -
          Rat Genome 230 2.0 Array 1394854_at - -
          Affymetrix HT Rat Focus Array 1394854_PM_at - -
          Affymetrix HT Rat Focus Array 1394854_at - -
          Affymetrix Rat Clariom S Assay PSR0500016295.rn.1 TC0500002133.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016296.rn.1 TC0500002133.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016294.rn.1 TC0500002133.rn.1 RTA TC
          Rat Expression Set 230 1394854_at - -
          Rat Expression Set 230 1394854_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10874940 - -
          Rat Genome U34 Set rc_AI045394_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21125523 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367102 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline