Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01749717_m1
          See other ZBTB8A GE Assays ›
          Gene Symbol:
          Zbtb8a
          Gene Name:
          zinc finger and BTB domain containing 8a
          Gene Aliases:
          Zbtb8
          Chromosome Location:
          Chr.5: 147554907 - 147584281 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01749717_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb8a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107913.1 NP_001101383.1 4 - 5 1323 73
          XM_006238931.2 XP_006238993.1 4 - 5 1739 73
          GenBank mRNA BC161892.1 - 4 - 5 1256 7191093 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003668.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          molecular_function
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P288982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598437 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177926 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139728 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P288982 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P288982 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P288982 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288982 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880000 - -
          Affymetrix Rat Gene 1.0 ST Array 10880003 - -
          Affymetrix Rat Gene 1.0 ST Array 10880001 - -
          Affymetrix Rat Gene 1.0 ST Array 10880002 - -
          Affymetrix Rat Gene 1.1 ST Array 10872312 - -
          Affymetrix Rat Gene 1.1 ST Array 10879999 - -
          Rat Genome 230 2.0 Array 1385472_at - -
          Rat Genome 230 2.0 Array 1397882_at - -
          Rat Genome 230 2.0 Array 1385472_PM_at - -
          Rat Genome 230 2.0 Array 1397882_PM_at - -
          Affymetrix HT Rat Focus Array 1397882_PM_at - -
          Affymetrix HT Rat Focus Array 1385472_PM_at - -
          Affymetrix HT Rat Focus Array 1397882_at - -
          Affymetrix HT Rat Focus Array 1385472_at - -
          Affymetrix Rat Clariom S Assay PSR0500027327.rn.1 TC0500003668.rn.1 RTA TC
          Rat Expression Set 230 1397882_PM_at - -
          Rat Expression Set 230 1397882_at - -
          Rat Expression Set 230 1385472_at - -
          Rat Expression Set 230 1385472_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10879999 - -
          Rat Genome U34 Set rc_AA963583_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22189765 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline