Search
Search
| Gene Symbol: Dhdds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011978.1 | NP_001011978.1 | 4 - 5 | 453 | 60 | |
| XM_006239014.2 | XP_006239076.1 | 4 - 5 | 568 | 60 | ||
| GenBank mRNA | BC085875.1 | - | 4 - 5 | 453 | 7191470 | 60 |
Set Membership: |
|