Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01749774_m1
          See other PITHD1 GE Assays ›
          Gene Symbol:
          Pithd1
          Gene Name:
          PITH (C-terminal proteasome-interacting domain of thioredoxin-like) domain containing 1
          Gene Aliases:
          RGD1308048
          Chromosome Location:
          Chr.5: 154322562 - 154333095 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01749774_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pithd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_216543.7 XP_216543.5 5 - 6 664 84

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          oxidoreductase

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P16575 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16575 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16575 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P16575 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P114729 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16575 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880662 - -
          Affymetrix Rat Gene 1.0 ST Array 10880660 - -
          Affymetrix Rat Gene 1.0 ST Array 10880661 - -
          Affymetrix Rat Gene 1.0 ST Array 10880664 - -
          Affymetrix Rat Gene 1.0 ST Array 10880663 - -
          Affymetrix Rat Gene 1.0 ST Array 10880659 - -
          Rat Genome 230 2.0 Array 1375354_at - -
          Rat Genome 230 2.0 Array 1374554_at - -
          Rat Genome 230 2.0 Array 1375354_PM_at - -
          Rat Genome 230 2.0 Array 1374554_PM_at - -
          Affymetrix HT Rat Focus Array 1374554_at - -
          Affymetrix HT Rat Focus Array 1374554_PM_at - -
          Rat Expression Set 230 1375354_PM_at - -
          Rat Expression Set 230 1374554_at - -
          Rat Expression Set 230 1374554_PM_at - -
          Rat Expression Set 230 1375354_at - -
          Affymetrix Rat Gene 1.0 ST Array 10880658 - -
          Rat Genome U34 Set rc_AA964241_at - -
          Rat Genome U34 Set rc_AA850529_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21342499 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356820 - -
          RatRef-12 Expression BeadChip ILMN_50988 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline