Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01750192_m1
          See other KCNG1 GE Assays ›
          Gene Symbol:
          Kcng1
          Gene Name:
          potassium channel, voltage-gated modifier subfamily G, member 1
          Gene Aliases:
          Kh2, Kv6.1
          Chromosome Location:
          Chr.3: 165020067 - 165039707 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01750192_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcng1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106545.1 NP_001100015.1 2 - 3 1192 87
          XM_008762484.1 XP_008760706.1 2 - 3 1011 87
          GenBank mRNA M81784.1 - 2 - 3 1017 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004844.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of ion transmembrane transport
          protein homooligomerization
          potassium ion transmembrane transport
          delayed rectifier potassium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P097372 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P097376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391225 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79638 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178253 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49490 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097376 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097372 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097372 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097376 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P097376 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P097372 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P215575 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852012 - -
          Affymetrix Rat Gene 1.0 ST Array 10852013 - -
          Affymetrix Rat Gene 1.0 ST Array 10852008 - -
          Affymetrix Rat Gene 1.0 ST Array 10852011 - -
          Affymetrix Rat Gene 1.0 ST Array 10852010 - -
          Affymetrix Rat Gene 1.0 ST Array 10852009 - -
          Affymetrix Rat Gene 1.1 ST Array 10852007 - -
          Affymetrix Rat Clariom S Assay PSR0300037162.rn.1 TC0300004844.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300037164.rn.1 TC0300004844.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300037161.rn.1 TC0300004844.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10852007 - -
          Rat Genome U34 Set M81784_at - -
          Rat Genome U34 Set M81784_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20846842 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362431 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline