Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01750347_m1
          See other TUBD1 GE Assays ›
          Gene Symbol:
          Tubd1
          Gene Name:
          tubulin, delta 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 73865828 - 73886976 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01750347_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tubd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105826.1 NP_001099296.1 8 - 9 1438 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001312.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          tubulin

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based process
          metabolic process
          protein polymerization
          GTPase activity
          structural constituent of cytoskeleton
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57988 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127096 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038142 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038142 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038142 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P352172 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94059 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10737390 - -
          Affymetrix Rat Gene 1.0 ST Array 10737393 - -
          Affymetrix Rat Gene 1.0 ST Array 10737392 - -
          Affymetrix Rat Gene 1.0 ST Array 10737395 - -
          Affymetrix Rat Gene 1.0 ST Array 10737394 - -
          Affymetrix Rat Gene 1.0 ST Array 10737391 - -
          Affymetrix Rat Gene 1.1 ST Array 10737389 - -
          Affymetrix Rat Clariom S Assay PSR1000012027.rn.1 TC1000001312.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000012028.rn.1 TC1000001312.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10737389 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21986045 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369024 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline