Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01750515_m1
          See other KCTD5 GE Assays ›
          Gene Symbol:
          Kctd5
          Gene Name:
          potassium channel tetramerization domain containing 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 13420154 - 13446135 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01750515_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kctd5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105768.1 NP_001099238.1 5 - 6 687 73
          GenBank mRNA BC168712.1 - 5 - 6 690 8366833 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002377.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Ion Channel 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein homooligomerization
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P916510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35285 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158062 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P916510 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P916510 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P916510 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P916510 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P356812 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37451 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83616 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15892 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10740965 - -
          Affymetrix Rat Gene 1.0 ST Array 10740961 - -
          Affymetrix Rat Gene 1.0 ST Array 10740964 - -
          Affymetrix Rat Gene 1.0 ST Array 10740963 - -
          Affymetrix Rat Gene 1.0 ST Array 10740962 - -
          Affymetrix Rat Gene 1.0 ST Array 10740960 - -
          Affymetrix Rat Gene 1.1 ST Array 10740959 - -
          Rat Genome 230 2.0 Array 1397764_at - -
          Rat Genome 230 2.0 Array 1397764_PM_at - -
          Rat Genome 230 2.0 Array 1392292_at - -
          Rat Genome 230 2.0 Array 1392292_PM_at - -
          Affymetrix HT Rat Focus Array 1392292_at - -
          Affymetrix HT Rat Focus Array 1392292_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000020947.rn.1 TC1000002377.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000020946.rn.1 TC1000002377.rn.1 RTA TC
          Rat Expression Set 230 1397764_at - -
          Rat Expression Set 230 1392292_at - -
          Rat Expression Set 230 1397764_PM_at - -
          Rat Expression Set 230 1392292_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10740959 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20859967 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372575 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline