Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01750528_m1
          See other SLC36A3 GE Assays ›
          Gene Symbol:
          Slc36a3
          Gene Name:
          solute carrier family 36, member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 40462752 - 40490083 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01750528_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc36a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024764.1 NP_001019935.1 9 - 10 1202 67
          XM_006246381.2 XP_006246443.1 8 - 9 1026 67
          XM_008767685.1 XP_008765907.1 10 - 11 1182 67
          GenBank mRNA BC097463.1 - 9 - 10 1202 7457579 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002822.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc36a3

          Panther Classification:

          Molecular Function -

          amino acid transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          L-alanine transport
          glycine transport
          proton transport
          proline transmembrane transport
          hydrogen ion transmembrane transport
          hydrogen:amino acid symporter activity
          hydrogen ion transmembrane transporter activity
          L-alanine transmembrane transporter activity
          glycine transmembrane transporter activity
          L-proline transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P222936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607238 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173539 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106922 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P222936 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P222936 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P222936 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P222936 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98805 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65660 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10742764 - -
          Affymetrix Rat Gene 1.0 ST Array 10742763 - -
          Affymetrix Rat Gene 1.0 ST Array 10742765 - -
          Affymetrix Rat Gene 1.0 ST Array 10742760 - -
          Affymetrix Rat Gene 1.0 ST Array 10742757 - -
          Affymetrix Rat Gene 1.0 ST Array 10742754 - -
          Affymetrix Rat Gene 1.0 ST Array 10742755 - -
          Affymetrix Rat Gene 1.0 ST Array 10742753 - -
          Affymetrix Rat Gene 1.0 ST Array 10742758 - -
          Affymetrix Rat Gene 1.0 ST Array 10742756 - -
          Affymetrix Rat Gene 1.0 ST Array 10742759 - -
          Affymetrix Rat Gene 1.0 ST Array 10742761 - -
          Affymetrix Rat Gene 1.1 ST Array 10742752 - -
          Rat Genome 230 2.0 Array 1385435_PM_at - -
          Rat Genome 230 2.0 Array 1385435_at - -
          Affymetrix HT Rat Focus Array 1385435_at - -
          Affymetrix HT Rat Focus Array 1385435_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000024773.rn.1 TC1000002822.rn.1 RTA TC
          Rat Expression Set 230 1385435_PM_at - -
          Rat Expression Set 230 1385435_at - -
          Affymetrix Rat Gene 1.0 ST Array 10742752 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362211 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530488 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline