Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01750800_m1
          See other DGKE GE Assays ›
          Gene Symbol:
          Dgke
          Gene Name:
          diacylglycerol kinase, epsilon 64kDa
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 76382630 - 76408224 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01750800_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dgke
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039341.1 NP_001034430.1 3 - 4 904 73
          XM_006247070.2 XP_006247132.1 3 - 4 1139 73
          XM_006247071.2 XP_006247133.1 3 - 4 1139 73
          GenBank mRNA BC110049.1 - 3 - 4 904 7316890 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003529.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          kinase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein kinase C-activating G-protein coupled receptor signaling pathway
          phosphorylation
          intracellular signal transduction
          NAD+ kinase activity
          diacylglycerol kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P105412 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P762828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469678 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107530 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105412 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P762828 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105412 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P762828 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P762828 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P105412 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P762828 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83798 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91813 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10746168 - -
          Affymetrix Rat Gene 1.0 ST Array 10746171 - -
          Affymetrix Rat Gene 1.0 ST Array 10746170 - -
          Affymetrix Rat Gene 1.0 ST Array 10746162 - -
          Affymetrix Rat Gene 1.0 ST Array 10746163 - -
          Affymetrix Rat Gene 1.0 ST Array 10746160 - -
          Affymetrix Rat Gene 1.0 ST Array 10746164 - -
          Affymetrix Rat Gene 1.0 ST Array 10746166 - -
          Affymetrix Rat Gene 1.0 ST Array 10746165 - -
          Affymetrix Rat Gene 1.0 ST Array 10746159 - -
          Affymetrix Rat Gene 1.0 ST Array 10746169 - -
          Affymetrix Rat Gene 1.0 ST Array 10746161 - -
          Affymetrix Rat Gene 1.0 ST Array 10746167 - -
          Affymetrix Rat Gene 1.1 ST Array 10746158 - -
          Rat Genome 230 2.0 Array 1397439_PM_at - -
          Rat Genome 230 2.0 Array 1397439_at - -
          Affymetrix Rat Clariom S Assay PSR1000031561.rn.1 TC1000003529.rn.1 RTA TC
          Rat Expression Set 230 1397439_PM_at - -
          Rat Expression Set 230 1397439_at - -
          Affymetrix Rat Gene 1.0 ST Array 10746158 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21462331 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360422 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline