Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01751085_m1
          See other TNS4 GE Assays ›
          Gene Symbol:
          Tns4
          Gene Name:
          tensin 4
          Gene Aliases:
          RGD1310402
          Chromosome Location:
          Chr.10: 86977504 - 87001256 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01751085_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tns4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024881.1 NP_001020052.1 9 - 10 1866 60
          GenBank mRNA BC097378.1 - 9 - 10 1866 7445075 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003728.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tns4

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          protein localization
          actin binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P043511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401933 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85488 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043511 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043511 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P043511 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P405843 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63155 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11655 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10747018 - -
          Affymetrix Rat Gene 1.0 ST Array 10747014 - -
          Affymetrix Rat Gene 1.0 ST Array 10747021 - -
          Affymetrix Rat Gene 1.0 ST Array 10747012 - -
          Affymetrix Rat Gene 1.0 ST Array 10747015 - -
          Affymetrix Rat Gene 1.0 ST Array 10747017 - -
          Affymetrix Rat Gene 1.0 ST Array 10747022 - -
          Affymetrix Rat Gene 1.0 ST Array 10747020 - -
          Affymetrix Rat Gene 1.0 ST Array 10747023 - -
          Affymetrix Rat Gene 1.0 ST Array 10747019 - -
          Affymetrix Rat Gene 1.0 ST Array 10747013 - -
          Affymetrix Rat Gene 1.0 ST Array 10747016 - -
          Affymetrix Rat Gene 1.1 ST Array 10747011 - -
          Rat Genome 230 2.0 Array 1378947_PM_at - -
          Rat Genome 230 2.0 Array 1378947_at - -
          Affymetrix HT Rat Focus Array 1378947_PM_at - -
          Affymetrix HT Rat Focus Array 1378947_at - -
          Affymetrix Rat Clariom S Assay PSR1000033389.rn.1 TC1000003728.rn.1 RTA TC
          Rat Expression Set 230 1378947_at - -
          Rat Expression Set 230 1378947_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10747011 - -
          Rat Genome U34 Set rc_AI179390_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362884 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline