Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01751213_m1
          See other CCNF GE Assays ›
          Gene Symbol:
          Ccnf
          Gene Name:
          cyclin F
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 13594687 - 13619935 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01751213_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccnf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100474.1 NP_001093944.1 5 - 6 575 82
          GenBank mRNA AF410816.1 - 5 - 6 575 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002385.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          kinase activator

          Gene Ontology Categories:

          Function(s)

          re-entry into mitotic cell cycle
          placenta development
          mitotic nuclear division
          negative regulation of centrosome duplication
          protein ubiquitination
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          cell division

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P025963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179545 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44746 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025963 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025963 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P025963 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15921 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P467269 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76360 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89033 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741040 - -
          Affymetrix Rat Gene 1.0 ST Array 10741029 - -
          Affymetrix Rat Gene 1.0 ST Array 10741036 - -
          Affymetrix Rat Gene 1.0 ST Array 10741031 - -
          Affymetrix Rat Gene 1.0 ST Array 10741042 - -
          Affymetrix Rat Gene 1.0 ST Array 10741034 - -
          Affymetrix Rat Gene 1.0 ST Array 10741030 - -
          Affymetrix Rat Gene 1.0 ST Array 10741037 - -
          Affymetrix Rat Gene 1.0 ST Array 10741038 - -
          Affymetrix Rat Gene 1.0 ST Array 10741035 - -
          Affymetrix Rat Gene 1.0 ST Array 10741039 - -
          Affymetrix Rat Gene 1.0 ST Array 10741041 - -
          Affymetrix Rat Gene 1.0 ST Array 10741032 - -
          Affymetrix Rat Gene 1.0 ST Array 10741033 - -
          Affymetrix Rat Gene 1.1 ST Array 10741028 - -
          Rat Genome 230 2.0 Array 1373468_at - -
          Rat Genome 230 2.0 Array 1382370_at - -
          Rat Genome 230 2.0 Array 1373468_PM_at - -
          Rat Genome 230 2.0 Array 1382370_PM_at - -
          Affymetrix HT Rat Focus Array 1382370_PM_at - -
          Affymetrix HT Rat Focus Array 1382370_at - -
          Affymetrix Rat Clariom S Assay PSR1000021102.rn.1 TC1000002385.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021101.rn.1 TC1000002385.rn.1 RTA TC
          Rat Expression Set 230 1382370_at - -
          Rat Expression Set 230 1382370_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741028 - -
          Rat Genome U34 Set rc_AA900901_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21996118 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352346 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline