Search
Search
| Gene Symbol: Rhot1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107026.2 | NP_001100496.1 | 13 - 14 | 1282 | 75 | |
| XM_006246985.2 | XP_006247047.1 | 13 - 14 | 1328 | 75 | ||
| XM_006246986.1 | XP_006247048.1 | 13 - 14 | 1328 | 75 | ||
| GenBank mRNA | BC129123.1 | - | 13 - 14 | 1282 | 7130774 | 75 |
| FQ211576.1 | - | 13 - 14 | 1230 | 75 | ||
Set Membership: |
|