Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01751559_m1
          See other LRRC48 GE Assays ›
          Gene Symbol:
          Lrrc48
          Gene Name:
          leucine rich repeat containing 48
          Gene Aliases:
          RGD1309150
          Chromosome Location:
          Chr.10: 46711518 - 46762261 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01751559_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc48
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013857.1 NP_001013879.1 6 - 7 833 60
          XM_006246462.2 XP_006246524.1 7 - 8 1101 60
          XM_006246463.2 XP_006246525.1 7 - 8 880 60
          XM_006246464.2 XP_006246526.1 7 - 8 1432 60
          XM_006246465.2 XP_006246527.1 7 - 8 993 60
          XM_008767782.1 XP_008766004.1 8 - 9 987 60
          XM_008767783.1 XP_008766005.1 7 - 8 2197 60
          XM_008767784.1 XP_008766006.1 8 - 9 2473 60
          XM_008767785.1 XP_008766007.1 7 - 8 880 60
          GenBank mRNA BC083838.1 - 6 - 7 833 7113397 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000749.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrrc48

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator phosphatase modulator

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P045661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822587 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150909 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95036 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46677 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045661 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045661 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P045661 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19941 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33009 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33308 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90728 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67431 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10734046 - -
          Affymetrix Rat Gene 1.0 ST Array 10734060 - -
          Affymetrix Rat Gene 1.0 ST Array 10734051 - -
          Affymetrix Rat Gene 1.0 ST Array 10734059 - -
          Affymetrix Rat Gene 1.0 ST Array 10734057 - -
          Affymetrix Rat Gene 1.0 ST Array 10734054 - -
          Affymetrix Rat Gene 1.0 ST Array 10734058 - -
          Affymetrix Rat Gene 1.0 ST Array 10734053 - -
          Affymetrix Rat Gene 1.0 ST Array 10734055 - -
          Affymetrix Rat Gene 1.0 ST Array 10734052 - -
          Affymetrix Rat Gene 1.0 ST Array 10734056 - -
          Affymetrix Rat Gene 1.0 ST Array 10734048 - -
          Affymetrix Rat Gene 1.0 ST Array 10734049 - -
          Affymetrix Rat Gene 1.0 ST Array 10734050 - -
          Affymetrix Rat Gene 1.0 ST Array 10734047 - -
          Affymetrix Rat Gene 1.1 ST Array 10734045 - -
          Rat Genome 230 2.0 Array 1391765_at - -
          Rat Genome 230 2.0 Array 1391765_PM_at - -
          Affymetrix HT Rat Focus Array 1391765_at - -
          Affymetrix HT Rat Focus Array 1391765_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000006252.rn.1 TC1000000749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000006250.rn.1 TC1000000749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000006243.rn.1 TC1000000749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000006244.rn.1 TC1000000749.rn.1 RTA TC
          Rat Expression Set 230 1391765_PM_at - -
          Rat Expression Set 230 1391765_at - -
          Affymetrix Rat Gene 1.0 ST Array 10734045 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20699401 - -
          ABI Rat Genome Survey Microarray 21102326 - -
          ABI Rat Genome Survey Microarray 21344405 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356648 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline