Search
Search

Tbc1d9b| Gene Symbol: Tbc1d9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108274.1 | NP_001101744.1 | 10 - 11 | 1858 | 59 | |
| XM_006246291.2 | XP_006246353.1 | 10 - 11 | 1885 | 59 | ||
| XM_006246294.2 | XP_006246356.1 | 10 - 11 | 1885 | 59 | ||
| XM_006246295.2 | XP_006246357.1 | 10 - 11 | 1885 | 59 | ||
| XM_008767690.1 | XP_008765912.1 | 10 - 11 | 1781 | 59 | ||
| GenBank mRNA | BC166483.1 | - | 10 - 11 | 1794 | 7099086 | 59 |
Set Membership: |
|