Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01752207_g1
          See other MGC95210 GE Assays ›
          Gene Symbol:
          MGC95210
          Gene Name:
          hypothetical LOC287798
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 102199837 - 102210769 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01752207_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MGC95210
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005532.3 NP_001005532.2 4 - 5 2149 74
          XM_006247676.2 XP_006247738.1 3 - 4 1780 74
          XM_008768322.1 XP_008766544.1 3 - 4 1780 74
          XM_008768323.1 XP_008766545.1 3 - 4 1650 74
          XM_008768324.1 XP_008766546.1 3 - 4 1652 74
          XM_008768325.1 XP_008766547.1 4 - 5 1724 74
          XM_008768326.1 XP_008766548.1 4 - 5 1780 74
          XM_008768327.1 XP_008766549.1 3 - 4 1564 74
          XM_008768328.1 XP_008766550.1 3 - 4 1780 74
          XM_008768329.1 XP_008766551.1 3 - 4 1780 74
          XM_008768330.1 XP_008766552.1 3 - 4 2861 74
          XM_008768331.1 XP_008766553.1 1 - 2 1486 74
          GenBank mRNA BC082057.1 - 3 - 4 1510 7132329 74
          BC088445.1 - 3 - 4 1769 7126436 74
          BC099187.1 - 4 - 5 2149 7383366 74

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_008768331.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MGC95210

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1011244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648195 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86070 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94568 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1011244 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1011244 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1011244 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1011244 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61786 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17282 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10739390 - -
          Affymetrix Rat Gene 1.0 ST Array 10739387 - -
          Affymetrix Rat Gene 1.0 ST Array 10739393 - -
          Affymetrix Rat Gene 1.0 ST Array 10739392 - -
          Affymetrix Rat Gene 1.0 ST Array 10739388 - -
          Affymetrix Rat Gene 1.0 ST Array 10739389 - -
          Affymetrix Rat Gene 1.0 ST Array 10748780 - -
          Affymetrix Rat Gene 1.0 ST Array 10739391 - -
          Affymetrix Rat Gene 1.0 ST Array 10739394 - -
          Affymetrix Rat Gene 1.0 ST Array 10748781 - -
          Affymetrix Rat Gene 1.1 ST Array 10739386 - -
          Affymetrix Rat Gene 1.1 ST Array 10748776 - -
          Rat Genome 230 2.0 Array 1381391_PM_at - -
          Rat Genome 230 2.0 Array 1381391_at - -
          Affymetrix HT Rat Focus Array 1381391_at - -
          Affymetrix HT Rat Focus Array 1381391_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000016993.rn.1 TC1000001888.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016991.rn.1 TC1000001888.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016990.rn.1 TC1000001888.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016992.rn.1 TC1000001888.rn.1 RTA TC
          Rat Expression Set 230 1381391_at - -
          Rat Expression Set 230 1381391_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10739386 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353322 - -
          RatRef-12 Expression BeadChip ILMN_58274 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline