Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01752603_m1
          See other HORMAD2 GE Assays ›
          Gene Symbol:
          Hormad2
          Gene Name:
          HORMA domain containing 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 84576857 - 84662239 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01752603_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hormad2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017501.1 NP_001017501.1 8 - 9 657 108
          XM_008770340.1 XP_008768562.1 8 - 9 752 108
          XM_008770341.1 XP_008768563.1 10 - 11 909 108
          XM_008770342.1 XP_008768564.1 9 - 10 826 108
          XM_008770343.1 XP_008768565.1 8 - 9 608 108
          XM_008770344.1 XP_008768566.1 8 - 9 749 108
          XM_008770345.1 XP_008768567.1 3 - 4 1382 108
          GenBank mRNA BC079253.1 - 8 - 9 657 7133170 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002147.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hormad2

          Gene Ontology Categories:

          Function(s)

          meiotic sister chromatid cohesion

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P667084 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P095810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641501 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56705 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1270 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172357 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52640 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151815 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095810 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P667084 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095810 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P667084 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P667084 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P095810 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P667084 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20266 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23840 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9813 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36668 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7300 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77074 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22133 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10778070 - -
          Affymetrix Rat Gene 1.0 ST Array 10778075 - -
          Affymetrix Rat Gene 1.0 ST Array 10778076 - -
          Affymetrix Rat Gene 1.0 ST Array 10778074 - -
          Affymetrix Rat Gene 1.0 ST Array 10778078 - -
          Affymetrix Rat Gene 1.0 ST Array 10778073 - -
          Affymetrix Rat Gene 1.0 ST Array 10778077 - -
          Affymetrix Rat Gene 1.0 ST Array 10778079 - -
          Affymetrix Rat Gene 1.1 ST Array 10778069 - -
          Affymetrix Rat Clariom S Assay PSR1400015817.rn.1 TC1400002147.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10778069 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21653821 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351015 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline