Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01752908_g1
          See other IL2RG GE Assays ›
          Gene Symbol:
          Il2rg
          Gene Name:
          interleukin 2 receptor, gamma
          Gene Aliases:
          Ab2-183, Cd132
          Chromosome Location:
          Chr.X: 71165378 - 71169038 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01752908_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il2rg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080889.1 NP_543165.1 7 - 8 935 86
          GenBank mRNA AY325195.1 - 763 86
          BC079343.1 - 7 - 8 935 7106866 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001947.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il2rg

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          regulation of gene expression
          positive regulation of gene expression
          cytokine-mediated signaling pathway
          positive regulation of CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation
          positive regulation of T cell differentiation in thymus
          positive regulation of B cell differentiation
          positive regulation of lymphocyte differentiation
          cytokine receptor activity
          cytokine binding
          interleukin-2 binding
          interleukin-7 binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P050497 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P039878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12800 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039878 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P050497 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P050497 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039878 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P039878 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P050497 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1005524 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4140 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10938660 - -
          Affymetrix Rat Gene 1.0 ST Array 10938663 - -
          Affymetrix Rat Gene 1.0 ST Array 10938669 - -
          Affymetrix Rat Gene 1.0 ST Array 10938661 - -
          Affymetrix Rat Gene 1.0 ST Array 10938667 - -
          Affymetrix Rat Gene 1.0 ST Array 10938664 - -
          Affymetrix Rat Gene 1.0 ST Array 10938665 - -
          Affymetrix Rat Gene 1.0 ST Array 10938666 - -
          Affymetrix Rat Gene 1.0 ST Array 10938662 - -
          Affymetrix Rat Gene 1.1 ST Array 10938654 - -
          Rat Genome 230 2.0 Array 1389092_at - -
          Rat Genome 230 2.0 Array 1389092_PM_at - -
          Affymetrix HT Rat Focus Array 1389092_PM_at - -
          Affymetrix HT Rat Focus Array 1389092_at - -
          Affymetrix Rat Clariom S Assay PSR0X00012334.rn.1 TC0X00001947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00012337.rn.1 TC0X00001947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00012336.rn.1 TC0X00001947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00012338.rn.1 TC0X00001947.rn.1 RTA TC
          Rat Expression Set 230 1389092_PM_at - -
          Rat Expression Set 230 1389092_at - -
          Affymetrix Rat Gene 1.0 ST Array 10938654 - -
          Rat Genome U34 Set rc_AI059518_at - -
          Rat Genome U34 Set rc_AI178808_at - -
          Rat Genome U34 Set rc_AI172304_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22129016 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651036 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0