Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753062_m1
          See other HRASLS GE Assays ›
          Gene Symbol:
          Hrasls
          Gene Name:
          HRAS-like suppressor
          Gene Aliases:
          A-C1
          Chromosome Location:
          Chr.11: 75125951 - 75145866 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01753062_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hrasls
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105871.1 NP_001099341.1 2 - 3 292 78
          XM_006248510.2 XP_006248572.1 2 - 3 271 78
          GenBank mRNA AB510983.1 - 1 - 2 142 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001824.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          peroxisome organization
          ether lipid metabolic process
          phospholipase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P834031 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P129273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104157 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129273 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P834031 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P834031 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129273 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P834031 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P129273 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1039394 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3106 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10754989 - -
          Affymetrix Rat Gene 1.0 ST Array 10754990 - -
          Affymetrix Rat Gene 1.0 ST Array 10754988 - -
          Affymetrix Rat Gene 1.1 ST Array 10754987 - -
          Rat Genome 230 2.0 Array 1393790_at - -
          Rat Genome 230 2.0 Array 1393084_PM_at - -
          Rat Genome 230 2.0 Array 1393084_at - -
          Rat Genome 230 2.0 Array 1393790_PM_at - -
          Affymetrix HT Rat Focus Array 1393084_PM_at - -
          Affymetrix HT Rat Focus Array 1393790_at - -
          Affymetrix HT Rat Focus Array 1393084_at - -
          Affymetrix HT Rat Focus Array 1393790_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100011600.rn.1 TC1100001824.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011601.rn.1 TC1100001824.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011603.rn.1 TC1100001824.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011602.rn.1 TC1100001824.rn.1 RTA TC
          Rat Expression Set 230 1393790_PM_at - -
          Rat Expression Set 230 1393084_PM_at - -
          Rat Expression Set 230 1393084_at - -
          Rat Expression Set 230 1393790_at - -
          Affymetrix Rat Gene 1.0 ST Array 10754987 - -
          Rat Genome U34 Set rc_AI172003_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22348858 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354959 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline