Search
Search
| Gene Symbol: Clybl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100685.1 | NP_001094155.1 | 5 - 6 | 648 | 98 | |
| XM_006252508.2 | XP_006252570.1 | 4 - 5 | 477 | 98 | ||
| GenBank mRNA | BC088243.1 | - | 5 - 6 | 628 | 98 | |
| FQ214276.1 | - | 4 - 5 | 433 | 98 | ||
Set Membership: |
|