Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753636_m1
          See other POFUT1 GE Assays ›
          Gene Symbol:
          Pofut1
          Gene Name:
          protein O-fucosyltransferase 1
          Gene Aliases:
          Fut12
          Chromosome Location:
          Chr.3: 148722812 - 148749024 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01753636_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pofut1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001002278.1 NP_001002278.1 6 - 7 1002 84
          XM_006235290.2 XP_006235352.2 6 - 7 1146 84
          GenBank mRNA AJ781499.1 - 6 - 7 1002 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002061.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pofut1

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          somitogenesis
          fucose metabolic process
          protein O-linked glycosylation
          Notch signaling pathway
          nervous system development
          heart development
          biological_process
          protein O-linked fucosylation
          molecular_function
          fucosyltransferase activity
          peptide-O-fucosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P236025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657791 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17793 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175207 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48019 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P236025 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P236025 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P236025 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P236025 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66426 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92723 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10841015 - -
          Affymetrix Rat Gene 1.0 ST Array 10841018 - -
          Affymetrix Rat Gene 1.0 ST Array 10841014 - -
          Affymetrix Rat Gene 1.0 ST Array 10841013 - -
          Affymetrix Rat Gene 1.0 ST Array 10841019 - -
          Affymetrix Rat Gene 1.0 ST Array 10841017 - -
          Affymetrix Rat Gene 1.0 ST Array 10841012 - -
          Affymetrix Rat Gene 1.1 ST Array 10841011 - -
          Affymetrix Rat Clariom S Assay PSR0300014825.rn.1 TC0300002061.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10841011 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21182496 - -
          ABI Rat Genome Survey Microarray 22020836 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376278 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline