Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753837_m1
          See other ORC4 GE Assays ›
          Gene Symbol:
          Orc4
          Gene Name:
          origin recognition complex, subunit 4
          Gene Aliases:
          Orc4l
          Chromosome Location:
          Chr.3: 33035233 - 33075704 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01753837_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Orc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199092.1 NP_954523.1 13 - 14 1235 101
          XM_006234150.2 XP_006234212.1 15 - 16 1399 101
          XM_006234151.2 XP_006234213.1 14 - 15 1325 101
          XM_006234152.2 XP_006234214.1 16 - 17 1495 101
          XM_006234154.2 XP_006234216.1 12 - 13 1403 101
          XM_008761704.1 XP_008759926.1 13 - 14 1577 101
          XM_008761705.1 XP_008759927.1 15 - 16 1377 101
          XM_008761706.1 XP_008759928.1 14 - 15 1303 101
          XM_008761707.1 XP_008759929.1 14 - 15 1695 101
          XM_008761708.1 XP_008759930.1 13 - 14 1129 101
          GenBank mRNA BC060516.1 - 13 - 14 1235 5598062 101
          FQ213311.1 - 1185 101
          FQ226016.1 - 8 - 9 803 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003075.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Orc4

          Panther Classification:

          Molecular Function -

          replication origin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication initiation
          nucleotide binding
          DNA replication origin binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1018252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70649 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102715 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163599 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112042 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1018252 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1018252 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1018252 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1018252 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46474 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85865 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9326 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10845108 - -
          Affymetrix Rat Gene 1.0 ST Array 10845102 - -
          Affymetrix Rat Gene 1.0 ST Array 10845104 - -
          Affymetrix Rat Gene 1.0 ST Array 10845106 - -
          Affymetrix Rat Gene 1.0 ST Array 10845100 - -
          Affymetrix Rat Gene 1.0 ST Array 10845099 - -
          Affymetrix Rat Gene 1.0 ST Array 10845105 - -
          Affymetrix Rat Gene 1.0 ST Array 10845097 - -
          Affymetrix Rat Gene 1.0 ST Array 10845098 - -
          Affymetrix Rat Gene 1.0 ST Array 10845096 - -
          Affymetrix Rat Gene 1.0 ST Array 10845107 - -
          Affymetrix Rat Gene 1.0 ST Array 10845101 - -
          Affymetrix Rat Gene 1.0 ST Array 10845109 - -
          Affymetrix Rat Gene 1.0 ST Array 10845103 - -
          Affymetrix Rat Gene 1.1 ST Array 10845095 - -
          Rat Genome 230 2.0 Array 1391703_at - -
          Rat Genome 230 2.0 Array 1390478_at - -
          Rat Genome 230 2.0 Array 1390478_PM_at - -
          Rat Genome 230 2.0 Array 1391703_PM_at - -
          Affymetrix HT Rat Focus Array 1391703_PM_at - -
          Affymetrix HT Rat Focus Array 1391703_at - -
          Affymetrix Rat Clariom S Assay PSR0300023742.rn.1 TC0300003075.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300023741.rn.1 TC0300003075.rn.1 RTA TC
          Rat Expression Set 230 1390478_PM_at - -
          Rat Expression Set 230 1390478_at - -
          Rat Expression Set 230 1391703_PM_at - -
          Rat Expression Set 230 1391703_at - -
          Affymetrix Rat Gene 1.0 ST Array 10845095 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20875694 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353734 - -
          RatRef-12 Expression BeadChip ILMN_59082 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline