Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01754314_m1
          See other SRMS GE Assays ›
          Gene Symbol:
          Srms
          Gene Name:
          src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 176709742 - 176716146 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01754314_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Srms
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011961.1 NP_001011961.1 3 - 4 781 113
          GenBank mRNA BC090006.1 - 3 - 4 781 7323399 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300005047.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Srms

          Gene Ontology Categories:

          Function(s) Process(es)

          transmembrane receptor protein tyrosine kinase signaling pathway
          cell migration
          actin cytoskeleton organization
          cell differentiation
          peptidyl-tyrosine autophosphorylation
          regulation of cell proliferation
          innate immune response
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          receptor binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P044762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897091 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044762 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044762 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P044762 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P541426 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79203 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852490 - -
          Affymetrix Rat Gene 1.0 ST Array 10852485 - -
          Affymetrix Rat Gene 1.0 ST Array 10852487 - -
          Affymetrix Rat Gene 1.0 ST Array 10852484 - -
          Affymetrix Rat Gene 1.0 ST Array 10852491 - -
          Affymetrix Rat Gene 1.0 ST Array 10852489 - -
          Affymetrix Rat Gene 1.0 ST Array 10852488 - -
          Affymetrix Rat Gene 1.0 ST Array 10852486 - -
          Affymetrix Rat Gene 1.1 ST Array 10852483 - -
          Affymetrix Rat Clariom S Assay PSR0300038342.rn.1 TC0300005047.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10852483 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22113943 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376921 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline