Search
Search
| Gene Symbol: Rgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107299.1 | NP_001100769.1 | 3 - 4 | 397 | 82 | |
| XM_008771101.1 | XP_008769323.1 | 3 - 4 | 385 | 82 | ||
| XM_008771102.1 | XP_008769324.1 | 3 - 4 | 358 | 82 | ||
Set Membership: |
|