Search
Search
| Gene Symbol: Phf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108253.1 | NP_001101723.1 | 8 - 9 | 1181 | 65 | |
| XM_006256792.2 | XP_006256854.1 | 8 - 9 | 2013 | 65 | ||
| XM_006256794.2 | XP_006256856.1 | 8 - 9 | 1421 | 65 | ||
| XM_006256795.2 | XP_006256857.1 | 8 - 9 | 2013 | 65 | ||
| XM_006256796.2 | XP_006256858.1 | 8 - 9 | 2012 | 65 | ||
| XM_008773119.1 | XP_008771341.1 | 8 - 9 | 1151 | 65 | ||
Set Membership: |
|