Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01754888_m1
          See other RAB30 GE Assays ›
          Gene Symbol:
          Rab30
          Gene Name:
          RAB30, member RAS oncogene family
          Gene Aliases:
          Rsb30
          Chromosome Location:
          Chr.1: 157573344 - 157664297 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01754888_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab30
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001015012.1 NP_001015012.1 2 - 3 348 103
          XM_006229692.2 XP_006229754.1 4 - 5 573 103
          XM_006229693.2 XP_006229755.1 4 - 5 542 103
          XM_008759647.1 XP_008757869.1 3 - 4 3540 103
          GenBank mRNA BC091182.1 - 2 - 3 348 7326213 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002042.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab30

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          Golgi organization
          metabolic process
          Rab protein signal transduction
          lipid particle organization
          GTPase activity
          GTP binding
          GDP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P131086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600526 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103093 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P131086 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P131086 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P131086 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71015 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88960 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10708666 - -
          Affymetrix Rat Gene 1.0 ST Array 10708669 - -
          Affymetrix Rat Gene 1.0 ST Array 10708671 - -
          Affymetrix Rat Gene 1.0 ST Array 10708670 - -
          Affymetrix Rat Gene 1.0 ST Array 10708667 - -
          Affymetrix Rat Gene 1.1 ST Array 10708665 - -
          Rat Genome 230 2.0 Array 1379606_at - -
          Rat Genome 230 2.0 Array 1379606_PM_at - -
          Affymetrix HT Rat Focus Array 1379606_at - -
          Affymetrix HT Rat Focus Array 1379606_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100015719.rn.1 TC0100002042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100015723.rn.1 TC0100002042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100015724.rn.1 TC0100002042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100015722.rn.1 TC0100002042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100015725.rn.1 TC0100002042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100015718.rn.1 TC0100002042.rn.1 RTA TC
          Rat Expression Set 230 1379606_at - -
          Rat Expression Set 230 1379606_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10708665 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21095073 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361194 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline