Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01755060_m1
          See other CYB5R2 GE Assays ›
          Gene Symbol:
          Cyb5r2
          Gene Name:
          cytochrome b5 reductase 2
          Gene Aliases:
          RGD1308421
          Chromosome Location:
          Chr.1: 171971424 - 171980641 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01755060_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyb5r2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014244.1 NP_001014266.1 9 - 10 901 102
          XM_008759774.1 XP_008757996.1 7 - 8 2151 102
          XM_008759775.1 XP_008757997.1 8 - 9 2054 102
          XM_008759776.1 XP_008757998.1 8 - 9 2066 102
          XM_008759777.1 XP_008757999.1 7 - 8 2039 102
          XM_008759778.1 XP_008758000.1 8 - 9 1469 102
          XM_008759779.1 XP_008758001.1 8 - 9 1626 102
          XM_008759780.1 XP_008758002.1 6 - 7 655 102
          XM_008759781.1 XP_008758003.1 9 - 10 950 102
          XM_008759782.1 XP_008758004.1 7 - 8 2087 102
          GenBank mRNA BC079235.1 - 9 - 10 901 7132617 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006329.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyb5r2

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          sterol biosynthetic process
          oxidation-reduction process
          cytochrome-b5 reductase activity, acting on NAD(P)H

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P311597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67134 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55835 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P311597 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P311597 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P311597 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P311597 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27107 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55589 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724634 - -
          Affymetrix Rat Gene 1.0 ST Array 10724636 - -
          Affymetrix Rat Gene 1.0 ST Array 10724632 - -
          Affymetrix Rat Gene 1.0 ST Array 10724631 - -
          Affymetrix Rat Gene 1.0 ST Array 10724628 - -
          Affymetrix Rat Gene 1.0 ST Array 10724626 - -
          Affymetrix Rat Gene 1.0 ST Array 10724629 - -
          Affymetrix Rat Gene 1.0 ST Array 10724633 - -
          Affymetrix Rat Gene 1.0 ST Array 10724635 - -
          Affymetrix Rat Gene 1.0 ST Array 10724627 - -
          Affymetrix Rat Gene 1.1 ST Array 10724625 - -
          Rat Genome 230 2.0 Array 1391430_at - -
          Rat Genome 230 2.0 Array 1391430_PM_at - -
          Affymetrix HT Rat Focus Array 1391430_PM_at - -
          Affymetrix HT Rat Focus Array 1391430_at - -
          Affymetrix Rat Clariom S Assay PSR0100049839.rn.1 TC0100006329.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049840.rn.1 TC0100006329.rn.1 RTA TC
          Rat Expression Set 230 1391430_PM_at - -
          Rat Expression Set 230 1391430_at - -
          Affymetrix Rat Gene 1.0 ST Array 10724625 - -
          Rat Genome U34 Set rc_AI112240_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21870008 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373498 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline