Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01755118_g1
          See other HIRIP3 GE Assays ›
          Gene Symbol:
          Hirip3
          Gene Name:
          HIRA interacting protein 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 198298138 - 198301164 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01755118_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hirip3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025725.1 NP_001020896.1 7 - 8 1804 86
          XM_006230307.1 XP_006230369.1 6 - 7 1501 86
          GenBank mRNA BC099091.1 - 7 - 8 1804 7457888 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002608.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hirip3

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P129063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317665 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129063 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129063 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P129063 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P841993 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10710952 - -
          Affymetrix Rat Gene 1.0 ST Array 10710948 - -
          Affymetrix Rat Gene 1.0 ST Array 10710956 - -
          Affymetrix Rat Gene 1.0 ST Array 10710955 - -
          Affymetrix Rat Gene 1.0 ST Array 10710951 - -
          Affymetrix Rat Gene 1.0 ST Array 10710954 - -
          Affymetrix Rat Gene 1.0 ST Array 10710950 - -
          Affymetrix Rat Gene 1.0 ST Array 10710957 - -
          Affymetrix Rat Gene 1.1 ST Array 10710947 - -
          Rat Genome 230 2.0 Array 1371813_PM_at - -
          Rat Genome 230 2.0 Array 1371813_at - -
          Affymetrix HT Rat Focus Array 1371813_at - -
          Affymetrix HT Rat Focus Array 1371813_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100020240.rn.1 TC0100002608.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100020241.rn.1 TC0100002608.rn.1 RTA TC
          Rat Expression Set 230 1371813_at - -
          Rat Expression Set 230 1371813_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10710947 - -
          Rat Genome U34 Set rc_AI060150_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21222618 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355241 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline