Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01755844_g1
          See other PRR3 GE Assays ›
          Gene Symbol:
          Prr3
          Gene Name:
          proline rich 3
          Gene Aliases:
          Cat56
          Chromosome Location:
          Chr.20: 3300190 - 3307262 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01755844_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_212544.2 NP_997709.1 3 - 4 494 92
          XM_006255942.1 XP_006256004.1 4 - 5 562 92
          XM_006255943.2 XP_006256005.1 3 - 4 380 92
          GenBank mRNA BC098636.1 - 4 - 5 572 92
          BC126073.1 - 3 - 4 494 7311139 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000097.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prr3

          Gene Ontology Categories:

          Process(es)

          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P299349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75287 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P299349 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P299349 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P299349 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P299349 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10827845 - -
          Affymetrix Rat Gene 1.0 ST Array 10827842 - -
          Affymetrix Rat Gene 1.0 ST Array 10827843 - -
          Affymetrix Rat Gene 1.0 ST Array 10827844 - -
          Affymetrix Rat Gene 1.1 ST Array 10827840 - -
          Rat Genome 230 2.0 Array 1371603_at - -
          Rat Genome 230 2.0 Array 1371603_PM_at - -
          Affymetrix HT Rat Focus Array 1371603_PM_at - -
          Affymetrix HT Rat Focus Array 1371603_at - -
          Affymetrix Rat Clariom S Assay PSR2000000593.rn.1 TC2000000097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000594.rn.1 TC2000000097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000595.rn.1 TC2000000097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000596.rn.1 TC2000000097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000591.rn.1 TC2000000097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000597.rn.1 TC2000000097.rn.1 RTA TC
          Rat Expression Set 230 1371603_PM_at - -
          Rat Expression Set 230 1371603_at - -
          Affymetrix Rat Gene 1.0 ST Array 10827840 - -
          Rat Genome U34 Set rc_AI236705_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21994912 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361060 - -
          RatRef-12 Expression BeadChip ILMN_60410 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline