Search
Search

LOC102549842| Gene Symbol: LOC102549842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006227817.2 | XP_006227879.1 | 3 - 4 | 367 | 66 | |
| XM_008758701.1 | XP_008756923.1 | 3 - 4 | 1216 | 66 | ||
Set Membership: |
|