Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01756034_m1
          See other AKAP7 GE Assays ›
          Gene Symbol:
          Akap7
          Gene Name:
          A kinase (PRKA) anchor protein 7
          Gene Aliases:
          AKAP-18, AKAP18d, Akap15, Akap18
          Chromosome Location:
          Chr.1: 21129213 - 21263850 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01756034_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Akap7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001801.3 NP_001001801.1 2 - 3 311 96
          GenBank mRNA AY350741.1 - 2 - 3 311 96
          BC098632.1 - 428 7315112 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000224.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Akap7

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein localization
          regulation of protein kinase A signaling
          regulation of membrane repolarization
          cellular response to cAMP
          positive regulation of potassium ion transmembrane transport
          positive regulation of delayed rectifier potassium channel activity
          protein binding
          protein C-terminus binding
          AMP binding
          protein kinase binding
          protein domain specific binding
          protein complex scaffold
          protein kinase A regulatory subunit binding
          protein kinase A binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P004720 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P004725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798703 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62865 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153926 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111509 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115525 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124929 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152528 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58880 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134979 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103148 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137141 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166954 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163023 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26917 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15945 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76278 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004725 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P004725 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P228995 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P823597 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50001 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58045 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19356 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104629 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35216 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24301 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92359 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47212 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71406 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38791 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75291 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10702213 - -
          Affymetrix Rat Gene 1.0 ST Array 10702212 - -
          Affymetrix Rat Gene 1.0 ST Array 10702203 - -
          Affymetrix Rat Gene 1.0 ST Array 10702207 - -
          Affymetrix Rat Gene 1.0 ST Array 10702204 - -
          Affymetrix Rat Gene 1.0 ST Array 10702209 - -
          Affymetrix Rat Gene 1.0 ST Array 10702205 - -
          Affymetrix Rat Gene 1.0 ST Array 10702208 - -
          Affymetrix Rat Gene 1.0 ST Array 10702202 - -
          Affymetrix Rat Gene 1.0 ST Array 10702206 - -
          Affymetrix Rat Gene 1.1 ST Array 10702201 - -
          Rat Genome 230 2.0 Array 1389496_PM_at - -
          Rat Genome 230 2.0 Array 1389496_at - -
          Rat Genome 230 2.0 Array 1392079_at - -
          Rat Genome 230 2.0 Array 1392079_PM_at - -
          Affymetrix HT Rat Focus Array 1392079_at - -
          Affymetrix HT Rat Focus Array 1389496_PM_at - -
          Affymetrix HT Rat Focus Array 1392079_PM_at - -
          Affymetrix HT Rat Focus Array 1389496_at - -
          Affymetrix Rat Clariom S Assay PSR0100001239.rn.1 TC0100000224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001244.rn.1 TC0100000224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001243.rn.1 TC0100000224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001252.rn.1 TC0100000224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001240.rn.1 TC0100000224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001241.rn.1 TC0100000224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001250.rn.1 TC0100000224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001251.rn.1 TC0100000224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001238.rn.1 TC0100000224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001242.rn.1 TC0100000224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001235.rn.1 TC0100000224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001249.rn.1 TC0100000224.rn.1 RTA TC
          Rat Expression Set 230 1389496_at - -
          Rat Expression Set 230 1389496_PM_at - -
          Rat Expression Set 230 1392079_PM_at - -
          Rat Expression Set 230 1392079_at - -
          Affymetrix Rat Gene 1.0 ST Array 10702201 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22320727 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376565 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline