Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01756365_m1
          See other TMEM184A GE Assays ›
          Gene Symbol:
          Tmem184a
          Gene Name:
          transmembrane protein 184A
          Gene Aliases:
          RGD1306702
          Chromosome Location:
          Chr.12: 16906867 - 16922459 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01756365_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem184a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025413.1 NP_001020584.1 1 - 2 239 64
          XM_006248938.2 XP_006249000.1 2 - 3 435 64
          XM_006248939.2 XP_006249001.1 2 - 3 427 64
          XM_006248940.2 XP_006249002.1 2 - 3 435 64
          XM_008768972.1 XP_008767194.1 2 - 3 251 64
          GenBank mRNA BC083910.1 - 1 - 2 237 64
          BC098056.1 - 1 - 2 239 7457725 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000292.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem184a

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s)

          germ-line sex determination
          regulation of protein localization
          regulation of secretion

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729648 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63074 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011424 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011424 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011424 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P250433 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88569 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756999 - -
          Affymetrix Rat Gene 1.0 ST Array 10756994 - -
          Affymetrix Rat Gene 1.0 ST Array 10756997 - -
          Affymetrix Rat Gene 1.0 ST Array 10756993 - -
          Affymetrix Rat Gene 1.0 ST Array 10756995 - -
          Affymetrix Rat Gene 1.0 ST Array 10756998 - -
          Affymetrix Rat Gene 1.0 ST Array 10757000 - -
          Affymetrix Rat Gene 1.0 ST Array 10756996 - -
          Affymetrix Rat Gene 1.1 ST Array 10756991 - -
          Affymetrix Rat Clariom S Assay PSR1200002159.rn.1 TC1200000292.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200002157.rn.1 TC1200000292.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200002156.rn.1 TC1200000292.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200002160.rn.1 TC1200000292.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10756991 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22267496 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368827 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0