Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01756445_m1
          See other CYTH3 GE Assays ›
          Gene Symbol:
          Cyth3
          Gene Name:
          cytohesin 3
          Gene Aliases:
          Pscd3, Sec7, Sec7C
          Chromosome Location:
          Chr.12: 12877559 - 12907771 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01756445_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyth3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053912.2 NP_446364.2 7 - 8 831 86
          GenBank mRNA BC101884.1 - 7 - 8 831 7929513 86
          U83897.1 - 7 - 8 824 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001114.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyth3

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          vesicle-mediated transport
          regulation of cell adhesion
          regulation of ARF protein signal transduction
          positive regulation of GTPase activity
          positive regulation of cell adhesion
          Golgi vesicle transport
          establishment of epithelial cell polarity
          ARF guanyl-nucleotide exchange factor activity
          phosphatidylinositol-3,4,5-trisphosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P137168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883229 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168306 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84397 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63712 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137168 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137168 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P137168 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P299268 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54605 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37870 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5893 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10760389 - -
          Affymetrix Rat Gene 1.0 ST Array 10760381 - -
          Affymetrix Rat Gene 1.0 ST Array 10760390 - -
          Affymetrix Rat Gene 1.0 ST Array 10760383 - -
          Affymetrix Rat Gene 1.0 ST Array 10760385 - -
          Affymetrix Rat Gene 1.0 ST Array 10760384 - -
          Affymetrix Rat Gene 1.0 ST Array 10760386 - -
          Affymetrix Rat Gene 1.0 ST Array 10760382 - -
          Affymetrix Rat Gene 1.0 ST Array 10760392 - -
          Affymetrix Rat Gene 1.0 ST Array 10760391 - -
          Affymetrix Rat Gene 1.0 ST Array 10760387 - -
          Affymetrix Rat Gene 1.0 ST Array 10760380 - -
          Affymetrix Rat Gene 1.0 ST Array 10760388 - -
          Affymetrix Rat Gene 1.1 ST Array 10760379 - -
          Rat Genome 230 2.0 Array 1387513_PM_at - -
          Rat Genome 230 2.0 Array 1387513_at - -
          Rat Genome 230 2.0 Array 1388712_at - -
          Rat Genome 230 2.0 Array 1388712_PM_at - -
          Affymetrix HT Rat Focus Array 1387513_at - -
          Affymetrix HT Rat Focus Array 1388712_PM_at - -
          Affymetrix HT Rat Focus Array 1387513_PM_at - -
          Affymetrix HT Rat Focus Array 1388712_at - -
          Affymetrix Rat Clariom S Assay PSR1200009559.rn.1 TC1200001114.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200009560.rn.1 TC1200001114.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200009561.rn.1 TC1200001114.rn.1 RTA TC
          Rat Expression Set 230 1388712_at - -
          Rat Expression Set 230 1387513_at - -
          Rat Expression Set 230 1388712_PM_at - -
          Rat Expression Set 230 1387513_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10760379 - -
          Rat Genome U34 Set U83897_at - -
          Rat Genome U34 Set rc_AI176246_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21615407 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355570 - -
          RatRef-12 Expression BeadChip ILMN_64245 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline