Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01756450_m1
          See other PRKAR1B GE Assays ›
          Gene Symbol:
          Prkar1b
          Gene Name:
          protein kinase, cAMP dependent regulatory, type I, beta
          Gene Aliases:
          RGD1563094, RIbeta
          Chromosome Location:
          Chr.12: 17614472 - 17713567 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01756450_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prkar1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033679.1 NP_001028851.2 9 - 10 976 127
          XM_008768942.1 XP_008767164.1 8 - 9 1249 127
          GenBank mRNA BC091371.1 - 4 - 5 495 127
          BC104679.1 - 9 - 10 976 7456971 127
          FQ212815.1 - 8 - 9 1084 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000308.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prkar1b

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          learning or memory
          regulation of protein kinase activity
          negative regulation of cAMP-dependent protein kinase activity
          cAMP-dependent protein kinase inhibitor activity
          cAMP-dependent protein kinase regulator activity
          cAMP binding
          protein kinase A catalytic subunit binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P737148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250487 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160064 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63936 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50946 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108507 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4772 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53001 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46140 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14686 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117297 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66821 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P737148 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P737148 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P737148 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P170755 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29582 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64685 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69994 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62022 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99680 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43350 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72432 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73604 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757121 - -
          Affymetrix Rat Gene 1.0 ST Array 10757125 - -
          Affymetrix Rat Gene 1.0 ST Array 10757126 - -
          Affymetrix Rat Gene 1.0 ST Array 10757122 - -
          Affymetrix Rat Gene 1.0 ST Array 10757127 - -
          Affymetrix Rat Gene 1.0 ST Array 10757120 - -
          Affymetrix Rat Gene 1.0 ST Array 10757123 - -
          Affymetrix Rat Gene 1.0 ST Array 10757124 - -
          Affymetrix Rat Gene 1.0 ST Array 10757128 - -
          Affymetrix Rat Gene 1.1 ST Array 10757119 - -
          Rat Genome 230 2.0 Array 1389463_at - -
          Rat Genome 230 2.0 Array 1389463_PM_at - -
          Affymetrix HT Rat Focus Array 1389463_PM_at - -
          Affymetrix HT Rat Focus Array 1389463_at - -
          Affymetrix Rat Clariom S Assay PSR1200002377.rn.1 TC1200000308.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200002380.rn.1 TC1200000308.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200002378.rn.1 TC1200000308.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200002376.rn.1 TC1200000308.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200002379.rn.1 TC1200000308.rn.1 RTA TC
          Rat Expression Set 230 1389463_at - -
          Rat Expression Set 230 1389463_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757119 - -
          Rat Genome U34 Set rc_H33086_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21473458 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline