Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01756526_m1
          See other RGD1566386 GE Assays ›
          Gene Symbol:
          RGD1566386
          Gene Name:
          similar to Hypothetical protein A430033K04
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 18105821 - 18139420 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01756526_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1566386
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107127.2 NP_001100597.2 3 - 4 325 114
          XM_008768973.1 XP_008767195.1 3 - 4 329 114
          GenBank mRNA BC167756.1 - 3 - 4 325 7099544 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001210.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904677 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32030 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113718 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70457 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058251 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058251 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058251 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P536502 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24712 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10760752 - -
          Affymetrix Rat Gene 1.0 ST Array 10760751 - -
          Affymetrix Rat Gene 1.0 ST Array 10760750 - -
          Affymetrix Rat Gene 1.0 ST Array 10760753 - -
          Affymetrix Rat Gene 1.1 ST Array 10760749 - -
          Rat Genome 230 2.0 Array 1393382_at - -
          Rat Genome 230 2.0 Array 1393382_PM_at - -
          Affymetrix HT Rat Focus Array 1393382_at - -
          Affymetrix HT Rat Focus Array 1393382_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200010312.rn.1 TC1200001210.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010313.rn.1 TC1200001210.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010311.rn.1 TC1200001210.rn.1 RTA TC
          Rat Expression Set 230 1393382_PM_at - -
          Rat Expression Set 230 1393382_at - -
          Affymetrix Rat Gene 1.0 ST Array 10760749 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359772 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline