Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01756784_m1
          See other PROZ GE Assays ›
          Gene Symbol:
          Proz
          Gene Name:
          protein Z, vitamin K-dependent plasma glycoprotein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 81784348 - 81797901 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01756784_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Proz
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006253473.1 XP_006253535.1 7 - 8 831 59
          XM_006253474.1 XP_006253536.1 6 - 7 717 59
          GenBank mRNA BC160868.1 - 7 - 8 743 59
          FQ209855.1 - 7 - 8 745 59

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          blood coagulation
          serine-type endopeptidase activity
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P008517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187093 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49033 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64375 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P433713 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89342 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74220 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792815 - -
          Affymetrix Rat Gene 1.0 ST Array 10792818 - -
          Affymetrix Rat Gene 1.0 ST Array 10792817 - -
          Affymetrix Rat Gene 1.0 ST Array 10792814 - -
          Affymetrix Rat Gene 1.0 ST Array 10792812 - -
          Affymetrix Rat Gene 1.0 ST Array 10792816 - -
          Affymetrix Rat Gene 1.0 ST Array 10792813 - -
          Affymetrix Rat Gene 1.0 ST Array 10792819 - -
          Affymetrix Rat Gene 1.1 ST Array 10792811 - -
          Affymetrix Rat Gene 1.0 ST Array 10792811 - -
          Rat Genome U34 Set rc_AI058639_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21391629 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360563 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline