Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rftn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135011.1 | NP_001128483.1 | 3 - 4 | 484 | 85 | |
XM_008766876.1 | XP_008765098.1 | 3 - 4 | 1048 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |