Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01757216_g1
          See other MRPS10 GE Assays ›
          Gene Symbol:
          Mrps10
          Gene Name:
          mitochondrial ribosomal protein S10
          Gene Aliases:
          Ac1179
          Chromosome Location:
          Chr.9: 15637427 - 15651154 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01757216_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrps10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008859.1 NP_001008859.1 5 - 6 420 71
          XM_006244464.2 XP_006244526.1 6 - 7 569 71
          XM_006244465.2 XP_006244527.1 6 - 7 566 71
          GenBank mRNA AY310148.1 - 5 - 6 420 71

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrps10

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P288015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101570 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118509 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P288015 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P288015 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P288015 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288015 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93154 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96822 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10926343 - -
          Affymetrix Rat Gene 1.0 ST Array 10926349 - -
          Affymetrix Rat Gene 1.0 ST Array 10926345 - -
          Affymetrix Rat Gene 1.0 ST Array 10926344 - -
          Affymetrix Rat Gene 1.0 ST Array 10926350 - -
          Affymetrix Rat Gene 1.0 ST Array 10926346 - -
          Affymetrix Rat Gene 1.1 ST Array 10926342 - -
          Affymetrix Rat Clariom S Assay PSR0900012837.rn.1 TC0900001544.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10926342 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21600070 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376421 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline